Tisk…

1-(4-Fluorophenyl)-1,3-butanedione 97%

Dodavatel: Apollo Scientific

Synonyms: (4-Fluorobenzoyl)acetone, 1-(4-Fluorophenyl)butane-1,3-dione

PC911392-5G
APOSPC911392-5GEA 2980 CZK
APOSPC911392-5G
1-(4-Fluorophenyl)-1,3-butanedione 97%
1-(4-Fluorophenyl)-1,3-butanedione
Vzorec: C₁₀H₉FO₂
MW: 180,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463139
CAS číslo: 29681-98-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR