Tisk…

3-Amino-6,7-difluoro-1H-indazole 95+%

Dodavatel: Apollo Scientific
Varování

PC911322-1G PC911322-250MG PC911322-5G
APOSPC911322-1GEA 1490 CZK
APOSPC911322-1G APOSPC911322-250MG APOSPC911322-5G
3-Amino-6,7-difluoro-1H-indazole 95+%
3-Amino-6,7-difluoro-1H-indazole
Vzorec: C₇H₅F₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD13191632
CAS číslo: 706805-37-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR