3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid
PC9112-25G
APOSPC9112-25GEA
15910
CZK
APOSPC9112-25G
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid 97%
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
Vzorec:
C₄H₂F₆O₂ MW: 196,048 g/mol Bod varu: 125 °C Bod tání: 46…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00464165 CAS číslo: 564-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...