(6-Fluoro-1H-benzimidazol-2-yl)methanol 95%
Dodavatel: Apollo Scientific
Synonyms:
(5-Fluoro-1H-benzo[d]imidazol-2-yl)methanol
PC911136-5G
APOSPC911136-5GEA
5790
CZK
APOSPC911136-5G
(6-Fluoro-1H-benzimidazol-2-yl)methanol 95%
(6-Fluoro-1H-benzimidazol-2-yl)methanol
Vzorec:
C₈H₇FN₂O MW: 166,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09261475 CAS číslo: 39811-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...