4-Fluoro-m-anisidine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Fluoro-3-anisidine, 4-Fluoro-3-methoxyaniline, 5-Amino-2-fluoroanisole
PC9111-25G
PC9111-5G
APOSPC9111-25GEA
1700
CZK
APOSPC9111-25G
APOSPC9111-5G
4-Fluoro-m-anisidine
4-Fluoro-m-anisidine
Vzorec:
C₇H₈FNO MW: 141,15 g/mol Bod tání: 66…68 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00665789 CAS číslo: 64465-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...