2-Amino-5-fluoro-3-methylpyridine 98%
Dodavatel: Apollo Scientific
Synonyms:
5-Fluoro-3-methylpyridin-2-ylamine
PC911092-1G
APOSPC911092-1GEA
1870
CZK
APOSPC911092-1G
2-Amino-5-fluoro-3-methylpyridine 98%
2-Amino-5-fluoro-3-methylpyridine
Vzorec:
C₆H₇FN₂ MW: 126,1300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07375130 CAS číslo: 886365-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...