Tisk…

2,2,2-Trifluoro-1-(pyridin-3-yl)ethanamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC911029-1G PC911029-500MG
APOSPC911029-1GEA 31910 CZK
APOSPC911029-1G APOSPC911029-500MG
2,2,2-Trifluoro-1-(pyridin-3-yl)ethanamine 97%
2,2,2-Trifluoro-1-(pyridin-3-yl)ethanamine
Vzorec: C₇H₇F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD07384536
CAS číslo: 912761-24-1
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR