Tisk…

5-Chloro-2-fluoroanisole 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Chloro-1-fluoro-2-methoxybenzene

PC910089-25G PC910089-5G
APOSPC910089-25GEA 8100 CZK
APOSPC910089-25G APOSPC910089-5G
5-Chloro-2-fluoroanisole 95%
5-Chloro-2-fluoroanisole
Vzorec: C₇H₆ClFO
MW: 160,58 g/mol
Storage Temperature: Ambient
CAS číslo: 1092349-89-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR