N-(Trifluoromethylthio)phthalimide
Dodavatel: Apollo Scientific
|
Varování
|
PC909739-1G
PC909739-5G
APOSPC909739-1GEA
6620
CZK
APOSPC909739-1G
APOSPC909739-5G
N-(Trifluoromethylthio)phthalimide
N-(Trifluoromethylthio)phthalimide
Vzorec:
C₉H₄F₃NO₂S Bod tání: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD18449627 CAS číslo: 719-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...