2-Fluoro-5-iodobenzaldehyde 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC9095-1G
PC9095-25G
PC9095-5G
APOSPC9095-1GEA
622
CZK
APOSPC9095-1G
APOSPC9095-25G
APOSPC9095-5G
2-Fluoro-5-iodobenzaldehyde 97%
2-Fluoro-5-iodobenzaldehyde
Vzorec:
C₇H₄FIO Bod tání: 41…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093999 CAS číslo: 146137-76-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...