Tisk…

2-Fluoro-4-iodobenzaldehyde

Dodavatel: Apollo Scientific
Varování

PC9094-1G PC9094-5G PC9094-25G
APOSPC9094-1GEA 1740 CZK
APOSPC9094-1G APOSPC9094-5G APOSPC9094-25G
2-Fluoro-4-iodobenzaldehyde
2-Fluoro-4-iodobenzaldehyde
Vzorec: C₇H₄FIO
MW: 250,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782056
CAS číslo: 699016-40-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR