Tisk…

Potassium isopropyltrifluoroborate 95%

Dodavatel: Apollo Scientific
Varování

PC909044-1G PC909044-25G PC909044-5G
APOSPC909044-1GEA 612 CZK
APOSPC909044-1G APOSPC909044-25G APOSPC909044-5G
Potassium isopropyltrifluoroborate 95%
Potassium isopropyltrifluoroborate
Vzorec: C₃H₇BF₃K
Storage Temperature: Freezer
MDL Number: MFCD04112718
CAS číslo: 1041642-13-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR