Tisk…

3-Fluoro-4-piperidinone hydrochloride 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Fluoro-4-piperidone Hydrochloride

PC908720-1G PC908720-5G PC908720-10G
APOSPC908720-1GEA 3740 CZK
APOSPC908720-1G APOSPC908720-5G APOSPC908720-10G
3-Fluoro-4-piperidinone hydrochloride 95%
3-Fluoro-4-piperidinone hydrochloride
Vzorec: C₅H₉ClFNO
MW: 153,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11506659
CAS číslo: 1070896-59-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR