Tisk…

4-Fluoro-5-hydroxy-2-methylindole 95%

Dodavatel: Apollo Scientific

PC908657-1G PC908657-250MG
APOSPC908657-1GEA 1740 CZK
APOSPC908657-1G APOSPC908657-250MG
4-Fluoro-5-hydroxy-2-methylindole 95%
4-Fluoro-5-hydroxy-2-methylindole
Vzorec: C₉H₈FNO
MW: 165,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368140
CAS číslo: 288385-88-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR