Tisk…

3-Amino-2-(trifluoromethyl)pyridine 95%

Dodavatel: Apollo Scientific

PC908641-1G PC908641-250MG
APOSPC908641-1GEA 7110 CZK
APOSPC908641-1G APOSPC908641-250MG
3-Amino-2-(trifluoromethyl)pyridine 95%
3-Amino-2-(trifluoromethyl)pyridine
Vzorec: C₆H₅F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD09753488
CAS číslo: 106877-32-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR