Tisk…

Perfluorobutane-1-sulphonic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Perfluoro-1-butanesulphonic acid, Nonafluoro-1-butanesulphonic acid

PC9086-25G PC9086-5G
APOSPC9086-25GEA 4070 CZK
APOSPC9086-25G APOSPC9086-5G
Perfluorobutane-1-sulphonic acid 97%
Perfluorobutane-1-sulphonic acid

Nonaflates are slightly more reactive towards nucleophilic displacements than triflates.

Vzorec: C₄HF₉O₃S
Bod varu: 76…84 °C/0,1mmHg
Hustota: 1,811 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01320794
CAS číslo: 375-73-5
EINECS: 206-793-1
UN: 2586
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR