2-(Fluoromethyl)-6-formylpyridine 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-(Fluoromethyl)picolinaldehyde
PC908550-100MG
PC908550-250MG
APOSPC908550-100MGEA
870
CZK
APOSPC908550-100MG
APOSPC908550-250MG
2-(Fluoromethyl)-6-formylpyridine 95+%
2-(Fluoromethyl)-6-formylpyridine
Vzorec:
C₇H₆FNO MW: 139,1300 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11865234 CAS číslo: 208111-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...