2-Fluoro-3-methoxypyridine 95%
Dodavatel: Apollo Scientific
PC908496-1G
PC908496-5G
APOSPC908496-1GEA
857
CZK
APOSPC908496-1G
APOSPC908496-5G
2-Fluoro-3-methoxypyridine 95%
2-Fluoro-3-methoxypyridine
Vzorec:
C₆H₆FNO Storage Temperature: Ambient |
MDL Number:
MFCD09834126 CAS číslo: 163234-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...