Tisk…
2,3-Difluorophenyl isothiocyanate 95%
Dodavatel: Apollo Scientific
PC907988-1G PC907988-250MG
APOSPC907988-1GEA 16960 CZK
APOSPC907988-1G APOSPC907988-250MG
2,3-Difluorophenyl isothiocyanate 95%
2,3-Difluorophenyl isothiocyanate
Vzorec: C₇H₃F₂NS
Storage Temperature: Ambient
CAS číslo: 363179-57-1