Ethyl 4-Fluorocinnamate 95%
Dodavatel: Apollo Scientific
Synonyms:
4-Fluorocinnamic acid ethyl ester, Ethyl (2E)-3-(4-fluorophenyl)acrylate
PC907942-1G
PC907942-5G
APOSPC907942-1GEA
4370
CZK
APOSPC907942-1G
APOSPC907942-5G
Ethyl 4-Fluorocinnamate 95%
Ethyl 4-Fluorocinnamate
Vzorec:
C₁₁H₁₁FO₂ MW: 194.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02252626 CAS číslo: 352-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...