Tisk…

3-Amino-2-fluorobenzonitrile 96%

Dodavatel: Apollo Scientific

PC907815-1G PC907815-5G
APOSPC907815-1GEA 4570 CZK
APOSPC907815-1G APOSPC907815-5G
3-Amino-2-fluorobenzonitrile 96%
3-Amino-2-fluorobenzonitrile
Vzorec: C₇H₅FN₂
Storage Temperature: Ambient
MDL Number: MFCD08062434
CAS číslo: 873697-68-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR