4-Amino-3-chloro-2,5,6-trifluoropyridine 95%
Dodavatel: Apollo Scientific
PC907751-1G
PC907751-5G
APOSPC907751-1GEA
7050
CZK
APOSPC907751-1G
APOSPC907751-5G
4-Amino-3-chloro-2,5,6-trifluoropyridine 95%
4-Amino-3-chloro-2,5,6-trifluoropyridine
Vzorec:
C₅H₂ClF₃N₂ MW: 182,53 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010856 CAS číslo: 2693-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...