Tisk…

4-(4-Fluoro-3-methylphenyl)butyric acid 95%

PC907547-1G PC907547-5G
APOSPC907547-1GEA 4770 CZK
APOSPC907547-1G APOSPC907547-5G
4-(4-Fluoro-3-methylphenyl)butyric acid 95%
4-(4-Fluoro-3-methylphenyl)butyric acid
Vzorec: C₁₁H₁₃FO₂
MW: 196,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07790543
CAS číslo: 331-43-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR