Tisk…

4-Chloro-8-fluoroquinazoline 95%

Dodavatel: Apollo Scientific
Varování

PC907298-100MG PC907298-250MG
APOSPC907298-100MGEA 953 CZK
APOSPC907298-100MG APOSPC907298-250MG
4-Chloro-8-fluoroquinazoline 95%
4-Chloro-8-fluoroquinazoline
Vzorec: C₈H₄ClFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD09837193
CAS číslo: 124429-27-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR