Tisk…

2-Methyl-6-(trifluoromethyl)pyridine 95%

Dodavatel: Apollo Scientific

PC906598-5G
APOSPC906598-5GEA 3310 CZK
APOSPC906598-5G
2-Methyl-6-(trifluoromethyl)pyridine 95%
2-Methyl-6-(trifluoromethyl)pyridine
Vzorec: C₇H₆F₃N
Storage Temperature: Ambient
MDL Number: MFCD07774130
CAS číslo: 1620-72-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR