4-Fluorophenyl chlorothionoformate 98%
Dodavatel: Apollo Scientific
Synonyms:
O-(4-Fluorophenyl) Chlorothioformate, Chlorothioformic Acid O-(4-Fluorophenyl) Ester
PC906433-1G
PC906433-5G
APOSPC906433-1GEA
3110
CZK
APOSPC906433-1G
APOSPC906433-5G
4-Fluorophenyl chlorothionoformate 98%
4-Fluorophenyl chlorothionoformate
Vzorec:
C₇H₄ClFOS MW: 190,62 g/mol Bod varu: 82 °C Bod tání: 84 °C Hustota: 1,37 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134402 CAS číslo: 42908-73-6 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...