Tisk…

5-Fluoro-2-methylbenzoxazole 97%

Dodavatel: Apollo Scientific

Synonyms: 5-Fluoro-2-methylbenzo[d]oxazole

PC906427-5G
APOSPC906427-5GEA 3060 CZK
APOSPC906427-5G
5-Fluoro-2-methylbenzoxazole 97%
5-Fluoro-2-methylbenzoxazole
Vzorec: C₈H₆FNO
MW: 151,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093471
CAS číslo: 701-16-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR