Tisk…

3-(2-Fluorophenyl)propionaldehyde 95%

Dodavatel: Apollo Scientific

PC905581-1G PC905581-5G
APOSPC905581-1GEA 3380 CZK
APOSPC905581-1G APOSPC905581-5G
3-(2-Fluorophenyl)propionaldehyde 95%
3-(2-Fluorophenyl)propionaldehyde
Vzorec: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD09028588
CAS číslo: 175143-93-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR