Tisk…

(2-(Trifluoromethyl)pyridin-3-yl)methanol 95%

Dodavatel: Apollo Scientific

PC905183-1G PC905183-5G
APOSPC905183-1GEA 2940 CZK
APOSPC905183-1G APOSPC905183-5G
(2-(Trifluoromethyl)pyridin-3-yl)methanol 95%
(2-(Trifluoromethyl)pyridin-3-yl)methanol
Vzorec: C₇H₆F₃NO
Storage Temperature: Ambient
MDL Number: MFCD11100743
CAS číslo: 131747-57-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR