Tisk…

4-Chloro-7-fluoroquinazoline 95%

Dodavatel: Apollo Scientific

PC905039-5G PC905039-25G
APOSPC905039-5GEA 3580 CZK
APOSPC905039-5G APOSPC905039-25G
4-Chloro-7-fluoroquinazoline 95%
4-Chloro-7-fluoroquinazoline
Vzorec: C₈H₄ClFN₂
MW: 182.5800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236729
CAS číslo: 16499-62-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR