1-(2-Chloro-5-fluoropyridin-3-yl)ethanone 95%
Dodavatel: Apollo Scientific
PC904959-1G
PC904959-250MG
APOSPC904959-1GEA
3110
CZK
APOSPC904959-1G
APOSPC904959-250MG
1-(2-Chloro-5-fluoropyridin-3-yl)ethanone 95%
1-(2-Chloro-5-fluoropyridin-3-yl)ethanone
Vzorec:
C₇H₅ClFNO Storage Temperature: Ambient |
MDL Number:
MFCD13563063 CAS číslo: 1203499-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...