Tisk…

4-(4-Fluorophenyl)imidazole 98%

Dodavatel: Apollo Scientific

PC904946-10G PC904946-25G
APOSPC904946-10GEA 5390 CZK
APOSPC904946-10G APOSPC904946-25G
4-(4-Fluorophenyl)imidazole 98%
4-(4-Fluorophenyl)imidazole
Vzorec: C₉H₇FN₂
MW: 162,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00297041
CAS číslo: 65020-70-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR