Tisk…

1-(5-Fluoropyridin-2-yl)ethanamine 95%

Dodavatel: Apollo Scientific

PC904892-1G PC904892-250MG
APOSPC904892-1GEA 22950 CZK
APOSPC904892-1G APOSPC904892-250MG
1-(5-Fluoropyridin-2-yl)ethanamine 95%
1-(5-Fluoropyridin-2-yl)ethanamine
Vzorec: C₇H₉FN₂
Storage Temperature: Ambient
MDL Number: MFCD18260255
CAS číslo: 915720-57-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR