(4-Fluorophenyl)acetaldehyde 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(4-Fluorophenyl)acetaldehyde
PC904883-1G
PC904883-250MG
APOSPC904883-1GEA
9160
CZK
APOSPC904883-1G
APOSPC904883-250MG
(4-Fluorophenyl)acetaldehyde 95%
(4-Fluorophenyl)acetaldehyde
Vzorec:
C₈H₇FO MW: 138,14 g/mol Bod varu: 204 °C/760mmHg Storage Temperature: Ambient |
MDL Number:
MFCD02261761 CAS číslo: 1736-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...