5-Fluoro-2-(1H-pyrazol-1-yl)benzaldehyde 95%
Dodavatel: Apollo Scientific
PC904683-1G
PC904683-250MG
PC904683-5G
APOSPC904683-1GEA
6220
CZK
APOSPC904683-1G
APOSPC904683-250MG
APOSPC904683-5G
5-Fluoro-2-(1H-pyrazol-1-yl)benzaldehyde 95%
5-Fluoro-2-(1H-pyrazol-1-yl)benzaldehyde
Vzorec:
C₁₀H₇FN₂O Storage Temperature: Ambient |
MDL Number:
MFCD08059861 CAS číslo: 1015845-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...