Tisk…

3,3-Difluoro-1-boc-azetidine

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 3,3-difluoroazetidine-1-carboxylate

PC904450-5G PC904450-25G
APOSPC904450-5GEA 1780 CZK
APOSPC904450-5G APOSPC904450-25G
3,3-Difluoro-1-boc-azetidine
3,3-Difluoro-1-boc-azetidine
Vzorec: C₈H₁₃F₂NO₂
MW: 193,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18381087
CAS číslo: 1255666-59-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR