[5-(4-Fluorophenyl)-3-isoxazolyl]methanol 95%
Dodavatel: Apollo Scientific
PC904348-1G
PC904348-250MG
APOSPC904348-1GEA
4860
CZK
APOSPC904348-1G
APOSPC904348-250MG
[5-(4-Fluorophenyl)-3-isoxazolyl]methanol 95%
[5-(4-Fluorophenyl)-3-isoxazolyl]methanol
Vzorec:
C₁₀H₈FNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06798098 CAS číslo: 640291-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...