Tisk…

3,4-Difluorobenzoylacetonitrile 95+%

Dodavatel: Apollo Scientific

PC9042-1G PC9042-250MG PC9042-5G
APOSPC9042-1GEA 2240 CZK
APOSPC9042-1G APOSPC9042-250MG APOSPC9042-5G
3,4-Difluorobenzoylacetonitrile 95+%
3,4-Difluorobenzoylacetonitrile
Vzorec: C₉H₅F₂NO
MW: 181,14 g/mol
Storage Temperature: Ambient
CAS číslo: 71682-97-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR