Methyl 3,3-difluorocyclopentane-1-carboxylate 95%
Dodavatel: Apollo Scientific
PC904108-1G
PC904108-250MG
APOSPC904108-1GEA
18420
CZK
APOSPC904108-1G
APOSPC904108-250MG
Methyl 3,3-difluorocyclopentane-1-carboxylate 95%
Methyl 3,3-difluorocyclopentane-1-carboxylate
Vzorec:
C₇H₁₀F₂O₂ MW: 164,15 g/mol Storage Temperature: Ambient |
CAS číslo:
1394129-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...