Tisk…

6-Fluorochromane-2-carboxylic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 6-Fluorochroman-2-carboxylic acid, 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid

PC903709-25G
APOSPC903709-25GEA 3540 CZK
APOSPC903709-25G
6-Fluorochromane-2-carboxylic acid 98%
6-Fluorochromane-2-carboxylic acid
Vzorec: C₁₀H₉FO₃
MW: 196,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07778357
CAS číslo: 99199-60-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR