Ethyl-4-amino-2-fluorobenzoate 95%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 4-amino-2-fluorobenzoate
PC903639-5G
APOSPC903639-5GEA
3970
CZK
APOSPC903639-5G
Ethyl-4-amino-2-fluorobenzoate 95%
Ethyl-4-amino-2-fluorobenzoate
Vzorec:
C₉H₁₀FNO₂ MW: 183,1800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09037895 CAS číslo: 73792-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...