(2E)-4,4,4-Trifluoro-N-methoxy-N-methyl-2-butenamide 95%
Dodavatel: Apollo Scientific
PC903616-1G
PC903616-250MG
APOSPC903616-1GEA
12400
CZK
APOSPC903616-1G
APOSPC903616-250MG
(2E)-4,4,4-Trifluoro-N-methoxy-N-methyl-2-butenamide 95%
(2E)-4,4,4-Trifluoro-N-methoxy-N-methyl-2-butenamide
Vzorec:
C₆H₈F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22123285 CAS číslo: 1172603-97-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...