1-(Methoxymethyl)-4-(trifluoromethyl)benzene 96%
Dodavatel: Apollo Scientific
PC902960-1G
PC902960-5G
APOSPC902960-1GEA
2570
CZK
APOSPC902960-1G
APOSPC902960-5G
1-(Methoxymethyl)-4-(trifluoromethyl)benzene 96%
1-(Methoxymethyl)-4-(trifluoromethyl)benzene
Vzorec:
C₉H₉F₃O MW: 190,162 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11504959 CAS číslo: 155820-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...