N-(Chloroacetyl)-3-(trifluoromethyl)aniline
Dodavatel: Apollo Scientific
|
Varování
|
PC9028-25G
PC9028-100G
APOSPC9028-25GEA
4140
CZK
APOSPC9028-25G
APOSPC9028-100G
N-(Chloroacetyl)-3-(trifluoromethyl)aniline
N-(Chloroacetyl)-3-(trifluoromethyl)aniline
Vzorec:
C₉H₇ClF₃NO Storage Temperature: Ambient |
CAS číslo:
351-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...