Tisk…

2-Acetyl-3-fluoropyridine 98%

Dodavatel: Apollo Scientific
Varování

PC902771-1G PC902771-5G PC902771-25G
APOSPC902771-1GEA 662 CZK
APOSPC902771-1G APOSPC902771-5G APOSPC902771-25G
2-Acetyl-3-fluoropyridine 98%
2-Acetyl-3-fluoropyridine
Vzorec: C₇H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD06659495
CAS číslo: 87674-20-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR