5-(5-Fluoro-2-hydroxyphenyl)isoxazole 95%
Dodavatel: Apollo Scientific
Synonyms:
4-Fluoro-2-(5-isoxazolyl)phenol, 5-(5-Fluoro-2-hydroxyphenyl)isoxazole
PC902715-1G
PC902715-5G
APOSPC902715-1GEA
3110
CZK
APOSPC902715-1G
APOSPC902715-5G
5-(5-Fluoro-2-hydroxyphenyl)isoxazole 95%
5-(5-Fluoro-2-hydroxyphenyl)isoxazole
Vzorec:
C₉H₆FNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03070563 CAS číslo: 288401-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...