Tisk…

2-Fluoroethyl p-Toluenesulphonate 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Fluoroethyl Tosylate, 2-Fluoroethyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid 2-Fluoroethyl ester

PC902702-1G PC902702-25G PC902702-100G
APOSPC902702-1GEA 2700 CZK
APOSPC902702-1G APOSPC902702-25G APOSPC902702-100G
2-Fluoroethyl p-Toluenesulphonate 97%
2-Fluoroethyl p-Toluenesulphonate
Vzorec: C₉H₁₁FO₃S
MW: 218.24 g/mol
Hustota: 1.28 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01658061
CAS číslo: 383-50-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR