Tisk…

Ethyl-5-fluoro-1H-indole-2-carboxylate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 5-fluoro-1H-indole-2-carboxylate, Ethyl 5-Fluoroindole-2-carboxylate, 5-Fluoroindole-2-carboxylic acid ethyl ester

PC902680-5G PC902680-10G PC902680-25G
APOSPC902680-5GEA 4570 CZK
APOSPC902680-5G APOSPC902680-10G APOSPC902680-25G
Ethyl-5-fluoro-1H-indole-2-carboxylate 98%
Ethyl-5-fluoro-1H-indole-2-carboxylate
Vzorec: C₁₁H₁₀FNO₂
MW: 207,21 g/mol
Bod tání: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00152076
CAS číslo: 348-36-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR