7-Fluoro-1-methyl-1H-indazole 96%
Dodavatel: Apollo Scientific
PC902250-1G
PC902250-250MG
APOSPC902250-1GEA
6550
CZK
APOSPC902250-1G
APOSPC902250-250MG
7-Fluoro-1-methyl-1H-indazole 96%
7-Fluoro-1-methyl-1H-indazole
Vzorec:
C₈H₇FN₂ MW: 150.1500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12546530 CAS číslo: 1187386-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...