Tisk…

1-tert-Butyl-6-fluorobenzimidazole 98%

Dodavatel: Apollo Scientific

PC902196-1G PC902196-5G PC902196-25G
APOSPC902196-1GEA 3540 CZK
APOSPC902196-1G APOSPC902196-5G APOSPC902196-25G
1-tert-Butyl-6-fluorobenzimidazole 98%
1-tert-Butyl-6-fluorobenzimidazole
Vzorec: C₁₁H₁₃FN₂
Storage Temperature: Ambient
MDL Number: MFCD12546476
CAS číslo: 1187385-71-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR